Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014018.2 | NP_001014040.2 | 6 | 720 | ||
XM_008759352.1 | XP_008757574.1 | 5 | 2100 | |||
XM_008759353.1 | XP_008757575.1 | 6 | 2166 | |||
XM_008759354.1 | XP_008757576.1 | 6 | 1310 | |||
XM_008759355.1 | XP_008757577.1 | 7 | 1376 | |||
XM_008759356.1 | XP_008757578.1 | 6 | 1733 | |||
XM_008759357.1 | XP_008757579.1 | 6 | 1730 | |||
XM_008759358.1 | XP_008757580.1 | 6 | 1707 | |||
XM_008759359.1 | XP_008757581.1 | 6 | 1698 | |||
XM_008759360.1 | XP_008757582.1 | 6 | 938 | |||
XM_008759361.1 | XP_008757583.1 | 7 | 908 | |||
XM_008759362.1 | XP_008757584.1 | 7 | 836 | |||
XM_008759363.1 | XP_008757585.1 | 7 | 1004 | |||
XM_008759364.1 | XP_008757586.1 | 6 | 759 | |||
XM_008759365.1 | XP_008757587.1 | 7 | 827 | |||
GenBank mRNA | BC060592.1 | - | 6 | 724 | 6887297 | |
FQ220903.1 | - | 1 | 724 |