Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014018.2 | NP_001014040.2 | 6 | 1412 | ||
XM_008759352.1 | XP_008757574.1 | 5 | 2792 | |||
XM_008759353.1 | XP_008757575.1 | 6 | 2858 | |||
XM_008759354.1 | XP_008757576.1 | 6 | 2002 | |||
XM_008759355.1 | XP_008757577.1 | 7 | 2068 | |||
XM_008759356.1 | XP_008757578.1 | 6 | 2425 | |||
XM_008759357.1 | XP_008757579.1 | 6 | 2422 | |||
XM_008759358.1 | XP_008757580.1 | 6 | 2399 | |||
XM_008759359.1 | XP_008757581.1 | 6 | 2390 | |||
XM_008759360.1 | XP_008757582.1 | 6 | 1630 | |||
XM_008759361.1 | XP_008757583.1 | 7 | 1600 | |||
XM_008759362.1 | XP_008757584.1 | 7 | 1528 | |||
XM_008759363.1 | XP_008757585.1 | 7 | 1696 | |||
XM_008759364.1 | XP_008757586.1 | 6 | 1451 | |||
XM_008759365.1 | XP_008757587.1 | 7 | 1519 | |||
GenBank mRNA | BC060592.1 | - | 6 | 1416 | 6887297 | |
FM122595.1 | - | 1 | 364 | |||
FQ220903.1 | - | 1 | 1416 |