Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014023.1 | NP_001014045.1 | 8 | 2282 | ||
XM_006229912.2 | XP_006229974.1 | 8 | 2768 | |||
GenBank mRNA | BC078926.1 | - | 8 | 2282 | 7106563 | |
FQ225508.1 | - | 3 | 1402 | |||
FQ232280.1 | - | 1 | 1237 |