Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGOHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300739.2 | NP_001287668.1 | 2, 3 | 406 | ||
NM_001319985.1 | NP_001306914.1 | 2, 3 | 136 | |||
NM_018048.4 | NP_060518.1 | 2, 3 | 238 | |||
NR_135120.1 | 2, 3 | 238 | ||||
NR_135121.1 | 2, 3 | 136 | ||||
XM_005253402.1 | XP_005253459.1 | 3, 4 | 577 | |||
GenBank mRNA | AF165518.1 | - | 2, 3 | 234 | ||
AK001154.1 | - | 2, 3 | 223 | |||
AK022720.1 | - | 2, 3 | 213 | |||
AK096106.1 | - | 2, 3 | 229 | |||
BC010905.1 | - | 2, 3 | 201 | 4283108 | ||
BF667243.1 | - | 2, 3 | 118 | |||
BG497574.1 | - | 1, 2 | 69 | |||
BG533740.1 | - | 2, 3 | 239 | |||
BG539870.1 | - | 2, 3 | 119 | |||
BU933036.1 | - | 3, 4 | 457 | |||
CD357202.1 | - | 2, 3 | 241 | |||
KF051004.1 | - | 2, 3 | 161 |