Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmtr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014031.1 | NP_001014053.1 | 13 | 1562 | ||
XM_008772816.1 | XP_008771038.1 | 13 | 1632 | |||
GenBank mRNA | AY539880.1 | - | 1806 | |||
BC085799.1 | - | 13 | 1562 | 7110279 |