Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgms2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014043.1 | NP_001014065.1 | 3, 4 | 1010 | ||
XM_006233311.2 | XP_006233373.1 | 4, 5 | 1946 | |||
XM_008761501.1 | XP_008759723.1 | 3, 4 | 1862 | |||
GenBank mRNA | BC085803.1 | - | 3, 4 | 1010 | 7111151 | |
DQ071571.2 | - | 2, 3 | 581 |