Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sox5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271267.1 | NP_001258196.1 | 11 | 1554 | ||
XM_006237604.2 | XP_006237666.1 | 11 | 1603 | |||
XM_008763349.1 | XP_008761571.1 | 11 | 1574 | |||
XM_008763350.1 | XP_008761572.1 | 4 | 485 | |||
GenBank mRNA | BC086415.1 | - | 4 | 488 | 7131922 |