Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014065.2 | NP_001014087.1 | 5 | 723 | ||
XM_006257448.2 | XP_006257510.1 | 5 | 722 | |||
XM_006257449.2 | XP_006257511.1 | 5 | 733 | |||
XM_006257451.2 | XP_006257513.1 | 5 | 774 | |||
XM_006257452.2 | XP_006257514.1 | 4 | 705 | |||
XM_008773415.1 | XP_008771637.1 | 4 | 709 | |||
GenBank mRNA | BC089115.1 | - | 5 | 723 | 7319855 |