Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hpdl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014068.1 | NP_001014090.1 | 1 | 1556 | ||
GenBank mRNA | BC083702.1 | - | 1 | 1556 | 7190108 |