Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbcel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014089.1 | NP_001014111.1 | 8 | 1192 | ||
XM_006242859.2 | XP_006242921.1 | 8 | 3839 | |||
XM_008766134.1 | XP_008764356.1 | 9 | 3802 | |||
GenBank mRNA | BC087163.1 | - | 8 | 1192 | 7135124 |