Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paqr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014092.1 | NP_001014114.1 | 8 | 1392 | ||
GenBank mRNA | BC087040.1 | - | 8 | 1392 | 7106223 |
Set Membership: |
G-protein coupled receptor |