Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014098.1 | NP_001014120.1 | 4 | 1367 | ||
XM_006243788.2 | XP_006243850.1 | 4 | 1571 | |||
XM_006243789.2 | XP_006243851.1 | 4 | 2145 | |||
XM_006243790.2 | XP_006243852.1 | 4 | 1645 | |||
XM_006243791.2 | XP_006243853.1 | 4 | 1949 | |||
XM_006243792.2 | XP_006243854.1 | 4 | 1527 | |||
GenBank mRNA | BC087581.1 | - | 4 | 1367 | 7314505 |