Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018073.7 | NP_060543.5 | 2 | 555 | ||
GenBank mRNA | AF360739.1 | - | 2 | 508 | ||
AF439153.1 | - | 1 | 265 | |||
AK056523.1 | - | 2 | 517 | |||
AK291166.1 | - | 2 | 467 | |||
AK310888.1 | - | 2 | 464 | |||
AL832730.1 | - | 2 | 505 | |||
BC075058.2 | - | 1 | 289 | 30915367 | ||
BC109063.2 | - | 2 | 474 | 40033795 |