Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014135.1 | NP_001014157.1 | 15 | 2426 | ||
GenBank mRNA | BC085864.1 | - | 15 | 2426 | 7190485 |