Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014973.2 | NP_001014973.2 | 21 | 2368 | ||
GenBank mRNA | AK122352.1 | - | 10 | 1017 | ||
AK133353.1 | - | 18 | 1908 | |||
AK154425.1 | - | 21 | 2370 | |||
AK164161.1 | - | 21 | 2293 | |||
BC056394.1 | - | 21 | 2340 | 6852346 | ||
BC067201.1 | - | 21 | 2357 | 6825168 |