Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015006.1 | NP_001015006.1 | 10 | 1377 | ||
GenBank mRNA | BC092651.1 | - | 10 | 1377 | 7319534 |