Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfyve28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015039.1 | NP_001015039.1 | 10, 11 | 2562 | ||
GenBank mRNA | AK036334.1 | - | 10, 11 | 2566 | ||
AK129414.1 | - | 10, 11 | 2562 | |||
BC042774.1 | - | 3, 4 | 493 | |||
BC139050.1 | - | 10, 11 | 2503 | 8862072 | ||
BC139051.1 | - | 10, 11 | 2503 | 8862073 |