Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017453.1 | NP_001017453.1 | 6 | 1650 | ||
XM_006251293.2 | XP_006251355.1 | 6 | 2859 | |||
GenBank mRNA | BC092208.1 | - | 6 | 1650 | 7314987 | |
FQ219869.1 | - | 1 | 374 | |||
FQ229250.1 | - | 1 | 369 |