Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NECAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145277.1 | NP_001138749.1 | 2 | 251 | ||
NM_001145278.1 | NP_001138750.1 | 2 | 173 | |||
NM_018090.4 | NP_060560.1 | 2 | 251 | |||
XM_011541767.2 | XP_011540069.1 | 3 | 323 | |||
XM_017001721.1 | XP_016857210.1 | 2 | 199 | |||
GenBank mRNA | AK001282.1 | - | 2 | 183 | ||
AK023545.1 | - | 2 | 187 | |||
AK024468.1 | - | 2 | 165 | |||
AK223344.1 | - | 2 | 199 | |||
AK300764.1 | - | 2 | 197 | |||
AK302227.1 | - | 2 | 119 | |||
BC017014.1 | - | 2 | 182 | 3848563 | ||
BC018914.2 | - | 2 | 185 | 4299890 | ||
BM751287.1 | - | 2 | 251 |