Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA2026 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017969.2 | NP_001017969.2 | 8 | 5760 | ||
XM_005251385.2 | XP_005251442.1 | 7 | 6146 | |||
XM_006716730.3 | XP_006716793.1 | 7 | 3784 | |||
XM_011517761.2 | XP_011516063.1 | 5 | 3606 | |||
XM_017014332.1 | XP_016869821.1 | 4 | 3709 | |||
XM_017014333.1 | XP_016869822.1 | 5 | 3358 | |||
XM_017014334.1 | XP_016869823.1 | 4 | 3465 | |||
GenBank mRNA | AB095946.1 | - | 1 | 1809 | ||
AL833370.1 | - | 1 | 954 | |||
BC136846.1 | - | 3 | 3091 | 9020837 | ||
BX648372.1 | - | 4 | 3243 |