Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024255.1 | NP_001019426.1 | 14 | 1603 | ||
XM_008770141.1 | XP_008768363.1 | 13 | 1922 | |||
XM_008770142.1 | XP_008768364.1 | 13 | 1865 | |||
GenBank mRNA | BC095847.1 | - | 14 | 1603 | 7388955 |