Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024257.1 | NP_001019428.1 | 11 | 1281 | ||
XM_006255494.2 | XP_006255556.1 | 12 | 1399 | |||
XM_006255495.2 | XP_006255557.1 | 12 | 1358 | |||
GenBank mRNA | BC093396.1 | - | 11 | 1281 | 7388929 |