Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024773.1 | NP_001019944.1 | 4 | 1398 | ||
GenBank mRNA | BC097435.1 | - | 4 | 1398 | 7455454 |