Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024784.2 | NP_001019955.2 | 12 | 2221 | ||
XM_008763623.1 | XP_008761845.1 | 12 | 2439 | |||
GenBank mRNA | BC098037.1 | - | 12 | 2127 | 7456610 |