Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024786.1 | NP_001019957.1 | 16 | 2331 | ||
XM_008763872.1 | XP_008762094.1 | 15 | 2227 | |||
GenBank mRNA | BC097402.1 | - | 16 | 2331 | 7453940 |