Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sigirr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024887.1 | NP_001020058.1 | 10 | 1323 | ||
XM_006230552.2 | XP_006230614.1 | 10 | 2157 | |||
XM_006230553.2 | XP_006230615.1 | 10 | 1789 | |||
XM_008759988.1 | XP_008758210.1 | 9 | 3355 | |||
XM_008759989.1 | XP_008758211.1 | 10 | 1848 | |||
XM_008759990.1 | XP_008758212.1 | 10 | 1351 | |||
GenBank mRNA | BC097488.1 | - | 10 | 1323 | 7460323 |