Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321829.1 | NP_001308758.1 | 14 | 2521 | ||
NM_018017.3 | NP_060487.2 | 13 | 2396 | |||
NR_135815.1 | 14 | 3219 | ||||
XM_011539915.2 | XP_011538217.1 | 12 | 1739 | |||
GenBank mRNA | AF273054.1 | - | 12 | 1911 | ||
AK001050.1 | - | 3 | 290 | |||
AK057508.1 | - | 8 | 1014 | |||
AK307517.1 | - | 13 | 2373 | |||
AL834368.1 | - | 12 | 1595 | |||
BC022229.1 | - | 3 | 420 | |||
BC100670.1 | - | 14 | 2448 | |||
BC103499.1 | - | 13 | 2323 | 40001826 | ||
BC105632.1 | - | 13 | 2323 | 40001824 | ||
BC152306.1 | - | 13 | 2327 | 40012131 | ||
BX537964.1 | - | 13 | 2368 |