Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025121.1 | NP_001020292.1 | 8 | 935 | ||
XM_006240171.2 | XP_006240233.1 | 9 | 2053 | |||
XM_006240172.2 | XP_006240234.1 | 9 | 1200 | |||
XM_006240173.2 | XP_006240235.1 | 6 | 1458 | |||
GenBank mRNA | BC081896.1 | - | 8 | 935 | 7110321 |