Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGEH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014061.4 | NP_054780.2 | 1 | 549 | ||
GenBank mRNA | AF143235.4 | - | 1 | 545 | ||
AF320912.1 | - | 1 | 474 | |||
AK313527.1 | - | 1 | 473 | |||
BC011954.1 | - | 1 | 500 | 3880523 | ||
BI916425.1 | - | 1 | 549 |