Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006045.2 | NP_006036.1 | 28 | 5351 | ||
XM_011528481.2 | XP_011526783.1 | 28 | 5379 | |||
GenBank mRNA | AB014511.1 | - | 23 | 4907 | ||
AK025559.1 | - | 1 | 1179 | |||
AK026513.1 | - | 1 | 31 | |||
BC036759.1 | - | 1 | 1629 |