Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fpr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008042.2 | NP_032068.2 | 1 | 1029 | ||
GenBank mRNA | U78299.1 | - | 1077 |
Set Membership: |
G-protein coupled receptor |