Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm13242 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103158.2 | NP_001096628.2 | 5 | 3397 | ||
GenBank mRNA | BC096593.1 | - | 5 | 3409 | 30459933 |
Gene Symbol: Gm13247 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243138.1 | NP_001230067.1 | 5 | 2476 | ||
NM_001243139.1 | NP_001230068.1 | 4 | 2361 |
Gene Symbol: Rex2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177767.1 | NP_001171238.1 | 5 | 3398 | ||
GenBank mRNA | AK135667.1 | - | 79 | |||
BC067397.1 | - | 3 | 3153 | 30462971 |
Gene Symbol: Zfp600 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177545.2 | NP_001171016.1 | 5 | 3559 | ||
NM_001177546.2 | NP_001171017.1 | 5 | 3652 | |||
GenBank mRNA | BC108352.1 | - | 5 | 3559 | 30462028 |