Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrnp70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005045.2 | - | 2 | 329 | ||
AK011564.1 | - | 2 | 285 | |||
AK014311.2 | - | 2 | 326 | |||
AK077425.1 | - | 2 | 331 | |||
AK133115.1 | - | 2 | 330 | |||
AK146729.1 | - | 2 | 330 | |||
AK148013.1 | - | 2 | 356 | |||
BC049128.1 | - | 2 | 295 | |||
BC051414.1 | - | 1 | 130 | |||
BC131994.1 | - | 2 | 222 | 40130271 | ||
BC132020.1 | - | 2 | 222 | 40130297 |