Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112703.2 | NP_001106174.1 | 3 | 1639 | ||
NM_001283045.1 | NP_001269974.1 | 3 | 516 | |||
NM_001283046.1 | NP_001269975.1 | 4 | 1844 | |||
NM_001283047.1 | NP_001269976.1 | 4 | 1783 | |||
NM_009594.4 | NP_033724.2 | 3 | 410 | |||
GenBank mRNA | AK088252.1 | - | 4 | 1769 | ||
AK090095.1 | - | 3 | 368 | |||
AK166028.1 | - | 3 | 1628 | |||
AK166210.1 | - | 4 | 1878 | |||
BC059260.1 | - | 3 | 437 | 6408990 | ||
BC103770.1 | - | 3 | 863 | 30848553 | ||
CF536749.1 | - | 1 | 14 | |||
CJ099623.1 | - | 3 | 361 | |||
CJ128107.1 | - | 3 | 405 | |||
CN663033.1 | - | 2 | 131 | |||
J02995.1 | - | 3 | 363 | |||
K03228.1 | - | 3 | 405 | |||
M12263.1 | - | 4 | 770 | |||
M12264.1 | - | 3 | 516 | |||
M12265.1 | - | 3 | 565 |