Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRMD4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318336.1 | NP_001305265.1 | 3 | 566 | ||
NM_001318337.1 | NP_001305266.1 | 3 | 806 | |||
NM_018027.4 | NP_060497.3 | 4 | 506 | |||
XM_005252489.2 | XP_005252546.1 | 3 | 806 | |||
XM_005252490.2 | XP_005252547.1 | 3 | 566 | |||
XM_006717457.2 | XP_006717520.1 | 3 | 806 | |||
XM_006717458.3 | XP_006717521.1 | 3 | 806 | |||
XM_006717459.2 | XP_006717522.1 | 3 | 806 | |||
XM_006717460.2 | XP_006717523.1 | 3 | 806 | |||
XM_011519539.1 | XP_011517841.1 | 3 | 566 | |||
XM_011519540.1 | XP_011517842.1 | 3 | 806 | |||
XM_011519541.1 | XP_011517843.1 | 3 | 806 | |||
XM_011519543.2 | XP_011517845.1 | 4 | 574 | |||
XM_017016393.1 | XP_016871882.1 | 3 | 806 | |||
XM_017016394.1 | XP_016871883.1 | 4 | 574 | |||
XM_017016395.1 | XP_016871884.1 | 3 | 409 | |||
GenBank mRNA | AB037715.1 | - | 4 | 506 | ||
AK001072.1 | - | 3 | 574 | |||
AK094045.1 | - | 3 | 807 | |||
AK289693.1 | - | 4 | 431 | |||
BC151244.1 | - | 4 | 517 | 8860518 |