Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amelx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081978.2 | NP_001075447.1 | 5 | 235 | ||
NM_009666.4 | NP_033796.1 | 5 | 235 | |||
GenBank mRNA | AK029358.1 | - | 5 | 239 | ||
BC059090.1 | - | 5 | 205 | 30286944 | ||
CD773419.1 | - | 5 | 239 | |||
D31768.1 | - | 5 | 226 | |||
M10095.1 | - | 2 | 43 | |||
S74899.1 | - | 6 | 277 |