Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cux1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291233.1 | NP_001278162.1 | 2 | 645 | ||
NM_001291234.1 | NP_001278163.1 | 2 | 356 | |||
NM_001291238.1 | NP_001278167.1 | 2 | 356 | |||
NM_001291239.1 | NP_001278168.1 | 2 | 645 | |||
NM_001291240.1 | NP_001278169.1 | 2 | 645 | |||
NM_009986.4 | NP_034116.3 | 2 | 356 | |||
NM_198602.3 | NP_941004.2 | 2 | 356 | |||
GenBank mRNA | AB041595.1 | - | 2 | 492 | ||
AK098096.1 | - | 2 | 119 | |||
AK133543.1 | - | 2 | 119 | |||
AK143193.1 | - | 2 | 357 | |||
AK144932.1 | - | 2 | 1016 | |||
AK161760.1 | - | 2 | 130 | |||
AK220173.1 | - | 2 | 112 | |||
AY037807.1 | - | 2 | 86 | |||
BC014289.1 | - | 2 | 125 | |||
BC054370.1 | - | 2 | 79 | |||
BC079570.1 | - | 3 | 543 | |||
BC090847.1 | - | 2 | 128 | 30547458 | ||
CA317682.1 | - | 2 | 311 | |||
CJ177481.1 | - | 2 | 273 | |||
U68542.1 | - | 3 | 130 |