Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gng2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038637.1 | NP_001033726.1 | 5 | 2851 | ||
NM_001285908.1 | NP_001272837.1 | 4 | 2811 | |||
NM_001285909.1 | NP_001272838.1 | 6 | 2918 | |||
NM_001285910.1 | NP_001272839.1 | 3 | 2958 | |||
NM_001285911.1 | NP_001272840.1 | 4 | 2826 | |||
NM_010315.4 | NP_034445.1 | 4 | 2999 | |||
GenBank mRNA | AK003394.1 | - | 1 | 519 | ||
AK075698.1 | - | 4 | 2769 | |||
AK148899.1 | - | 1 | 1379 | |||
AK149133.1 | - | 1 | 927 | |||
AK158697.1 | - | 4 | 2998 | |||
AK160396.1 | - | 4 | 2727 |