Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Herc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010418.2 | NP_034548.2 | 30 | 4785 | ||
GenBank mRNA | AF061529.1 | - | 30 | 4735 | ||
AF071173.1 | - | 30 | 4687 | |||
AK140430.1 | - | 21 | 3410 | |||
AK141515.1 | - | 12 | 1760 |