Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ly6c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252055.1 | NP_001238984.1 | 3, 4 | 262 | ||
NM_001252056.1 | NP_001238985.1 | 3, 4 | 247 | |||
NM_010741.3 | NP_034871.2 | 3, 4 | 247 | |||
GenBank mRNA | AK088782.1 | - | 3, 4 | 224 | ||
AK143577.1 | - | 3, 4 | 239 | |||
BC010764.1 | - | 3, 4 | 240 | 4221342 | ||
BY118129.1 | - | 3, 4 | 221 | |||
CB595639.1 | - | 3, 4 | 250 | |||
CK031980.1 | - | 3, 4 | 213 |
Gene Symbol: Ly6c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099217.1 | NP_001092687.1 | 2, 3 | 204 | ||
GenBank mRNA | BC092082.1 | - | 2, 3 | 204 | 4983733 | |
M18466.1 | - | 2, 3 | 205 |