Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010886.2 | NP_035016.1 | 3 | 254 | ||
GenBank mRNA | AK005084.1 | - | 3 | 259 | ||
AK008357.1 | - | 3 | 251 | |||
AK008506.1 | - | 3 | 245 | |||
AK076036.1 | - | 3 | 255 | |||
AK154084.1 | - | 2 | 995 | |||
BC011114.1 | - | 3 | 217 | 3483028 | ||
U59509.1 | - | 3 | 152 |