Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRAMD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015124.4 | NP_055939.1 | 19 | 4298 | ||
XM_005261396.3 | XP_005261453.1 | 19 | 4234 | |||
XM_006724169.1 | XP_006724232.1 | 19 | 4196 | |||
XM_006724170.3 | XP_006724233.1 | 19 | 4215 | |||
XM_011530011.1 | XP_011528313.1 | 18 | 3705 | |||
XM_011530012.1 | XP_011528314.1 | 17 | 4065 | |||
XM_017028670.1 | XP_016884159.1 | 19 | 4256 | |||
GenBank mRNA | AB018310.2 | - | 19 | 4297 | ||
AL137628.1 | - | 1 | 1586 | |||
BC011984.2 | - | 1 | 941 | |||
BC063558.1 | - | 1 | 889 | |||
BC080189.1 | - | 4 | 2749 |