Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015135.2 | NP_055950.1 | 29 | 4131 | ||
XM_005250235.3 | XP_005250292.1 | 28 | 4083 | |||
GenBank mRNA | AK293497.1 | - | 7 | 918 | ||
BC044255.1 | - | 4 | 561 | |||
BC136624.1 | - | 29 | 4126 | 9020614 | ||
BC146784.1 | - | 29 | 4105 | 9007240 | ||
D86978.1 | - | 29 | 4105 |