Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syncrip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284328.1 | NP_001271257.1 | 6 | 927 | ||
NM_019666.2 | NP_062640.2 | 6 | 927 | |||
NM_019796.5 | NP_062770.1 | 6 | 927 | |||
GenBank mRNA | AB035725.1 | - | 6 | 726 | ||
AF093821.1 | - | 5 | 579 | |||
AF408434.1 | - | 4 | 380 | |||
AK034845.1 | - | 6 | 739 | |||
AK076026.1 | - | 6 | 719 | |||
AK077588.1 | - | 6 | 676 | |||
AK078158.1 | - | 4 | 360 | |||
AK083398.1 | - | 5 | 517 | |||
BC041148.1 | - | 6 | 772 | 4946652 | ||
BC055863.1 | - | 6 | 705 | |||
BC058807.1 | - | 6 | 794 | |||
BC080309.1 | - | 6 | 630 | 5716184 | ||
BC108363.1 | - | 6 | 661 | 4239809 | ||
CN529994.1 | - | 3 | 261 |