Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHJ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300836.1 | NP_001287765.1 | 4 | 364 | ||
NM_018049.2 | NP_060519.1 | 4 | 364 | |||
XM_017026924.1 | XP_016882413.1 | 4 | 687 | |||
XM_017026925.1 | XP_016882414.1 | 4 | 678 | |||
XM_017026926.1 | XP_016882415.1 | 4 | 687 | |||
XM_017026927.1 | XP_016882416.1 | 4 | 678 | |||
XM_017026928.1 | XP_016882417.1 | 4 | 678 | |||
GenBank mRNA | AK001159.1 | - | 4 | 340 | ||
AK092742.1 | - | 2 | 1889 | |||
AK097723.1 | - | 5 | 1371 | |||
AK098725.1 | - | 4 | 454 | |||
BC003084.2 | - | 4 | 285 | 2988404 | ||
BC008966.2 | - | 4 | 311 | 2961278 | ||
CN365243.1 | - | 4 | 283 | |||
CR457253.1 | - | 4 | 259 | |||
HY085955.1 | - | 4 | 354 | |||
HY101097.1 | - | 4 | 367 |