Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FOS SIRNA Assays ›
Gene Symbol:
Fos
Gene Name:
FBJ osteosarcoma oncogene
Gene Aliases:
c-fos
Chromosome Location:
Chr.6: 109300433 - 109303299 on Build Rnor_6.0
UniGene ID:
Rn.103750
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
261242
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Fos
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022197.2
NP_071533.1
1
237
GenBank mRNA
X06769.1
-
219
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_010234, AK154418, BC029814.
Panther Classification:
Molecular Function -
basic leucine zipper transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
conditioned taste aversion
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
transforming growth factor beta receptor signaling pathway
nervous system development
female pregnancy
aging
response to cold
response to light stimulus
response to mechanical stimulus
response to gravity
response to toxic substance
response to organic cyclic compound
sleep
cellular response to extracellular stimulus
response to lipopolysaccharide
response to progesterone
cellular response to hormone stimulus
response to cytokine
cellular response to reactive oxygen species
response to immobilization stress
skeletal muscle cell differentiation
response to muscle stretch
response to drug
positive regulation of osteoclast differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
response to corticosterone
response to cAMP
SMAD protein signal transduction
cellular response to calcium ion
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II core promoter sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
chromatin binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription factor binding
sequence-specific DNA binding
transcription regulatory region DNA binding
protein heterodimerization activity
R-SMAD binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-67fd5c5bdd-q6sk2:80/10.239.167.246:80.
git-commit: 7fb76fe7d0d673187a4c374cd16a9abd242c4866
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.3-offline