Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023046.4 | NP_075533.3 | 1, 2 | 104 | ||
GenBank mRNA | AK006515.1 | - | 1, 2 | 105 | ||
AK122456.1 | - | 1, 2 | 159 | |||
BC049699.1 | - | 1, 2 | 104 | |||
BU938650.1 | - | 1, 2 | 107 |