Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018052.3 | NP_060522.3 | 15 | 2074 | ||
XM_005256038.3 | XP_005256095.1 | 15 | 2085 | |||
GenBank mRNA | AK001167.1 | - | 5 | 533 | ||
AK025479.1 | - | 9 | 1107 | |||
AK056433.1 | - | 15 | 2084 | |||
AK093941.1 | - | 2 | 1053 | |||
AK297564.1 | - | 15 | 1859 | |||
AK307766.1 | - | 14 | 1924 | |||
AK310466.1 | - | 3 | 504 | |||
BC000536.2 | - | 5 | 538 | |||
BC007214.1 | - | 10 | 1196 | |||
BC110072.1 | - | 2 | 210 | |||
BC110852.1 | - | 10 | 1129 | |||
BC125108.1 | - | 15 | 2006 | 40118772 | ||
BC125109.1 | - | 15 | 2006 | 40118777 | ||
CR457343.1 | - | 9 | 1042 |