Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELOVL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104558.1 | NP_001098028.1 | 8 | 3044 | ||
NM_001297617.1 | NP_001284546.1 | 11 | 3442 | |||
NM_001297618.1 | NP_001284547.1 | 9 | 3049 | |||
NM_024930.2 | NP_079206.2 | 9 | 3095 | |||
XM_005248606.4 | XP_005248663.1 | 8 | 3659 | |||
XM_005248607.3 | XP_005248664.1 | 9 | 3372 | |||
XM_006714695.3 | XP_006714758.1 | 10 | 3151 | |||
XM_011543651.2 | XP_011541953.1 | 10 | 3341 | |||
XM_017009885.1 | XP_016865374.1 | 9 | 3032 | |||
XM_017009886.1 | XP_016865375.1 | 11 | 3271 | |||
XM_017009887.1 | XP_016865376.1 | 11 | 3462 | |||
GenBank mRNA | AB181393.1 | - | 9 | 3036 | ||
AL137506.1 | - | 4 | 2526 | |||
BC094792.1 | - | 11 | 3383 |