Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XKR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018053.3 | NP_060523.2 | 3 | 1874 | ||
XM_011541679.2 | XP_011539981.1 | 5 | 2145 | |||
XM_011541680.2 | XP_011539982.1 | 4 | 2037 | |||
GenBank mRNA | AK001169.1 | - | 1 | 541 | ||
AK026024.1 | - | 3 | 1343 | |||
AK054909.1 | - | 1 | 1967 | |||
AK091615.1 | - | 3 | 1874 | |||
AY534246.1 | - | 3 | 1332 | |||
BC003606.1 | - | 1 | 761 | |||
BC013379.2 | - | 3 | 1333 | 4025274 | ||
BC028564.2 | - | 3 | 1333 | 3832072 |
Gene Symbol: RNF216P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015449.1 | 6 | 1432 | |||
NR_023384.1 | 8 | 1605 | ||||
NR_023385.1 | 5 | 1317 | ||||
GenBank mRNA | AK124529.1 | - | 2 | 2810 | ||
AK308526.1 | - | 5 | 1252 | |||
BC026736.1 | - | 6 | 1311 | |||
BC068459.1 | - | 8 | 1574 | |||
BX538059.1 | - | 6 | 1359 |